Learn More
Invitrogen™ ACRBP Polyclonal Antibody

Description
Antibody detects endogenous levels of total ACRBP.
Specifications
Specifications
| Antigen | ACRBP |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ACRBP |
| Gene Accession No. | Q6AY33, Q8NEB7 |
| Gene Alias | ACRBP; acrbp {ECO:0000312; acrosin binding protein; acrosin-binding protein; Cancer/testis antigen 23; cancer/testis antigen OY-TES-1; CT23; EMBL:AAH79212.1}; OY-TES-1; proacrosin binding protein; proacrosin binding protein sp32; proacrosin-binding protein sp32; SP32; testicular tissue protein Li 10 |
| Gene Symbols | ACRBP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.