Learn More
Invitrogen™ ACRC Monoclonal Antibody (2E4)

Description
Peptide Sequence: HEMCHAASWL IDGIHDSHGD AWKYYARKSN RIHPELPRVT RCHNYKINYK VHYECTGCKT RIGCYTKSLD TSRFICAKCK GSLVMVPLTQ KDGTRIVPHV.
Specifications
Specifications
| Antigen | ACRC |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2E4 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | GCNA |
| Gene Accession No. | Q96QF7 |
| Gene Alias | acidic repeat containing; acidic repeat-containing protein; ACRC; GCNA; Germ cell nuclear antigen; NAAR1; putative nuclear protein |
| Gene Symbols | GCNA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.