Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACRC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554807
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554807 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554807 Supplier Invitrogen™ Supplier No. PA554807
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QSSSDDTSGS SVARRAPKRQ ASCILNVQSR SGDTSGSSVA RRAPKRQASS VVVIDSDSDE ECHTHEEKKA KLLEINSDDE SPECCHVKPA IQEPPIVISD DDNDDDNGND LE Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 26%.

Probable catalytic subunit of the acryloyl-CoA reductase complex involved in the pathway of L-alanine fermentation. Catalyzes the irreversible NADH-dependent formation of propionyl-CoA from acryloyl-CoA. It can also use 3-buten-2-one as substrate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACRC
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GCNA
Gene Accession No. Q96QF7
Gene Alias acidic repeat containing; acidic repeat-containing protein; ACRC; GCNA; Germ cell nuclear antigen; NAAR1; putative nuclear protein
Gene Symbols GCNA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACRC. Recombinant protein control fragment (Product #RP-91961).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93953
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.