Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acridine, 97%

Catalog No. aal0165706
Click to view available options
Quantity:
5 g
25 g

CAS: 260-94-6 | C13H9N | 179.222 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 260-94-6
Molecular Formula C13H9N
Molecular Weight (g/mol) 179.222
MDL Number MFCD00005025
InChI Key DZBUGLKDJFMEHC-UHFFFAOYSA-N
Synonym 9-azaanthracene, 2,3-benzoquinoline, acrydine, 10-azaanthracene, akridin, dibenzo b,e pyridine, 2,3,5,6-dibenzopyridine, akridin czech, benzo b quinoline
PubChem CID 9215
ChEBI CHEBI:36420
IUPAC Name acridine
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=N2

Specifications

Melting Point 107°C to 111°C
Density 1.005
Boiling Point 346°C
Quantity 5 g
UN Number UN2713
Beilstein 120200
Merck Index 14,122
Formula Weight 179.22
Percent Purity ≥98%
Chemical Name or Material Acridine
Hazard Category H302-H315-H319-H335-H340
Hazard Statement GHS H Statement
H341-H302-H315-H319-H335
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRIDINE
EINECSNumber 205-971-6
RTECSNumber AR7175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.