Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acridine, 98%, Thermo Scientific™

Catalog No. AC102760050
Change view
Click to view available options
:
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC102760050 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC102760050 Supplier Thermo Scientific Chemicals Supplier No. 102760050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 260-94-6
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C13H9N
MDL Number MFCD00005025
Beilstein 20, 459
Merck Index 15, 114
Synonym Dibenzo[b, e]pyridine
InChI Key DZBUGLKDJFMEHC-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=N2
Infrared Spectrum Authentic
IUPAC Name acridine
Molecular Weight (g/mol) 179.222
PubChem CID 9215
ChEBI CHEBI:36420
Formula Weight 179.22
Percent Purity ≥97.5% (HPLC)
Packaging Glass bottle
Color Brown-Yellow to Yellow
Quantity 5g
Chemical Name or Material Acridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause damage to organs.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.