Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Acridine Orange, pure, ca. 55% dye content

Catalog No. ac300910250
Click to view available options
Quantity:
25 g

CAS: 10127-02-3 | C17H20Cl3N3Zn | 438.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10127-02-3
Molecular Formula C17H20Cl3N3Zn
Molecular Weight (g/mol) 438.10
MDL Number MFCD00081043,MFCD00081043,MFCD00081043
InChI Key VADJQOXWNSPOQA-UHFFFAOYSA-L
Synonym 3,6-Bis(dimethylamino)acridine zinc chloride double salt, Basic Orange 14
PubChem CID 71774334
IUPAC Name zinc;hydron;3-N,3-N,6-N,6-N-tetramethylacridine-3,6-diamine;trichloride
SMILES [H+].[Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C

Specifications

Melting Point 165°C
UN Number 3143
Solubility Information Soluble in water
Formula Weight 369.94
Percent Purity 55%
Concentration or Composition (by Analyte or Components) N,N,N',N'-Tetramethylacridine-3,6-diamine monohydrochloride, compound with zinc dichloride
Odor Odorless
Color Index No. 46005
Color Brown/Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Acridine Orange
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed. Harmful if inhaled. Harmful in contact with skin. Suspected of causing genetic defects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Use personal protective equipment as required. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
DOTInformation Hazard Class: 6.1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.