Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acridine Yellow 98%, Thermo Scientific™

Catalog No. AC400240250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC400240250 25 g
AC400241000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC400240250 Supplier Thermo Scientific Chemicals Supplier No. 400240250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-49-9
Molecular Formula C15H15ClN3
Molecular Weight (g/mol) 272.76
MDL Number MFCD00012661
InChI Key MIVVOLJUKZMTIN-UHFFFAOYSA-N
Synonym 3, 6-Diamino-2, 7-dimethylacridine hydrochloride, C.I. 46025
PubChem CID 8672
ChEBI CHEBI:51742
IUPAC Name 2,7-dimethylacridine-3,6-diamine hydrochloridyl
SMILES [Cl].CC1=CC2=CC3=CC(C)=C(N)C=C3N=C2C=C1N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Beilstein 22, 488
Infrared Spectrum Authentic
Formula Weight 273.76
Percent Purity ≥97.5%
UV Spectrometry Complies
Color Brown/Red
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Acridine Yellow, 98%
Show More Show Less
EINECSNumber 205-194-2
RTECSNumber AR8790000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.