Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acriflavine Hydrochloride 90.0+%, TCI America™
SDP

Catalog No. A0677100G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
100 g

Chemical Identifiers

CAS 69235-50-3
Molecular Formula C27H27Cl3N6
Molecular Weight (g/mol) 541.91
MDL Number MFCD00069039
InChI Key MKLTXAHQKDVBLY-UHFFFAOYSA-N
Synonym unii-1s73vw819c, acrifiavine hydrochloride, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride
PubChem CID 53393742
IUPAC Name 3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine dihydrochloride chloride
SMILES Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12

Specifications

Quantity 100 g
UN Number 3077
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Acriflavine Hydrochloride (mixture of 3,6-Diamino-10-methylacridinium Chloride Hydrochloride and 3,6
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.