Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acriflavine neutral, 13.3-15.8% chlorine, ACROS Organics™
Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
Specifications
Packaging | Glass bottle |
Loss on Drying | 8% max. (105°C, 2 hrs) |
Melting Point | 179°C to 181°C |
Infrared Spectrum | Authentic |
Quantity | 100g |
Color | Brown-Red to Orange |
Beilstein | 22, I, 650 |
Merck Index | 15, 115 |
Solubility Information | Solubility in water: 1g/3mL. Other solubilities: incompletely soluble in alcohol,practically insoluble in ether,chloroform and,fixed oils |
Formula Weight | 469.03 |
Show More |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Warning
RTECSNumber : AR9660000