Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrolein Antibody (2H2), Novus Biologicals™
SDP

Catalog No. NBP259358 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259358 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259358 Supplier Novus Biologicals Supplier No. NBP259358
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Acrolein Monoclonal specifically detects Acrolein in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Acrolein
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2H2
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Flow Cytometry, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Formulation PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide
Gene Alias Acrylic amide; Ethylene Carboxamide 2-Propenamide; 2-Propen-1-one; 2-propenal; Acraldehyde; Acrolein; Acrylic aldehyde
Host Species Mouse
Immunogen Synthetic Acrolein modified Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for Acrolein modified proteins. Does not detect free acrolein. Does not cross-react with Crotonaldehyde, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins.
Target Species Non-species specific
Content And Storage Store at -20C.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.