Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrolein Mouse, PerCP, Clone: 2H2, Abnova™

Catalog No. 89338747 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-338-747 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-338-747 Supplier Abnova Corporation Supplier No. MAB19243
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

Acrolein Monoclonal antibody specifically detects Acrolein in samples. It is validated for WB, ELISA, ICC, IF, Flow Cyt.

Specifications

Antigen Acrolein
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone 2H2
Conjugate PerCP
Host Species Mouse
Immunogen Synthetic Acrolein modified Keyhole Limpet Hemocyanin (KLH)
Quantity 100 μg
Primary or Secondary Primary
Content And Storage Store at -20°C
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.