Learn More
Thermo Scientific™ AcroMetrix™ Adenovirus Controls (RUO)
Catalog No. 23279164
Change view
Click to view available options
Description:
AcroMetrix Adenovirus High Plasma Control
AcroMetrix Adenovirus Low Plasma Control
Level:
High
Low
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Level
23-279-164 AcroMetrix Adenovirus Low Plasma Control Low
23-279-165 AcroMetrix Adenovirus High Plasma Control High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-279-164 Supplier Thermo Scientific™ Supplier No. 954505
Add to Cart
Add to Cart
Identify increases in random or systematic error of nucleic acid test procedures for Adenovirus DNA with these controls, which are for Research Use Only.
Identify increases in random or systematic error of nucleic acid test procedures for the quantitative and qualitative determination of Adenovirus DNA with the Thermo Scientific™ AcroMetrix™ Adenovirus Controls (RUO). Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for Research Use Only and are not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY
Specifications
Description AcroMetrix Adenovirus Low Plasma Control
Type Adenovirus Control
Level Low
Packaging 5 x 0.5mL
Quantity 5 x 0.5 mL