Learn More
Thermo Scientific™ AcroMetrix™ CMV Controls (RUO)
Catalog No. 23279143
Change view
Click to view available options
Level:
High
Low
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
23-279-143 Low
23-279-144 High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-279-143 Supplier Thermo Scientific™ Supplier No. 951701
Add to Cart
Add to Cart
Identify increases in random or systematic error of nucleic acid test procedures for human cytomegalovirus (CMV) DNA with these controls, which are for Research Use Only.
Identify increases in random or systematic error of nucleic acid test procedures for the quantitative and qualitative determination of human cytomegalovirus (CMV) DNA with the Thermo Scientific™ AcroMetrix™ CMV Controls (RUO). Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for Research Use Only and are not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY
Specifications
Description AcroMetrix CMV Low Control
Type CMV Control
Level Low
Packaging 5 x 0.8mL
Quantity 5 x 0.8 mL