Learn More
Thermo Scientific™ AcroMetrix™ HBV Controls
Catalog No. 23279121
Change view
Click to view available options
Level:
High
Low
Mid
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
23-279-121 Mid
23-279-120 Low
23-279-122 High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-279-121 Supplier Thermo Scientific™ Supplier No. 965002
Add to Cart
Add to Cart
Identify increases in random or systematic error of nucleic acid tests for hepatitis B viral DNA (HBV DNA) with these controls, which are for In Vitro Diagnostic Use.
Identify increases in random or systematic error of nucleic acid test procedures for the quantitative and qualitative determination of hepatitis B viral DNA (HBV DNA) with the Thermo Scientific™ AcroMetrix™ HBV Controls. Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for In Vitro Diagnostic Use.
TRUSTED_SUSTAINABILITY
Specifications
Description AcroMetrix HBV Mid Control
Detectable Analytes HBV
Type HBV Control
Level Mid
Packaging 5 x 1mL
Quantity 5 x 1 mL