Learn More
Thermo Scientific™ AcroMetrix™ HCV Controls

Description
Monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation with the Thermo Scientific™ AcroMetrix™ HCV Controls. These controls identify random or systematic errors in nucleic acid test procedures for the quantitative and qualitative determination of hepatitis C viral RNA (HCV RNA) and can assist in identifying increases in random or systematic error. The AcroMetrix HCV Controls are not intended for use with assays used to screen the blood supply. They are for In Vitro Diagnostic Use.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | AcroMetrix HCV Mid Control |
| Detectable Analytes | HCV |
| Type | HCV Control |
| Level | Mid |
| Packaging | 5 x 1.2mL |
| Quantity | 5 x 1.2 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.