Learn More
Thermo Scientific™ AcroMetrix™ Herpes Simplex Virus Controls (RUO)
Catalog No. 23279154
Change view
Click to view available options
Description:
AcroMetrix HHV Negative Control
AcroMetrix HSV-1 High CSF Control
AcroMetrix HSV-1 Low Control
AcroMetrix HSV-2 High CSF Control
AcroMetrix HSV-2 Low Control
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
23-279-154 AcroMetrix HHV Negative Control
23-279-152 AcroMetrix HSV-1 Low Control
23-279-156 AcroMetrix HSV-2 Low Control
23-279-157 AcroMetrix HSV-2 High CSF Control
23-279-153 AcroMetrix HSV-1 High CSF Control
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 23-279-154 Supplier Thermo Scientific™ Supplier No. 951300
Add to Cart
Add to Cart
Assess the performance of molecular diagnostic test procedures for human herpes viruses such as HSV-1 and 2 with these controls, which are for Research Use Only.
Assess the performance of nucleic acid test procedures for the quantitative and qualitative determination of Human Herpes Viruses (HHV) with the Thermo Scientific™ AcroMetrix™ Herpes Simplex Virus Controls (RUO). Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for Research Use Only and are not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY
Specifications
Description AcroMetrix HHV Negative Control
Type HHV Control
Packaging 5 x 0.5mL
Quantity 5 x 0.5 mL