Learn More
Thermo Scientific™ AcroMetrix™ HIV-1 Controls
GSA_VA
Catalog No. 23279116
Change view
Click to view available options
Description:
AcroMetrix HIV-1 High Control
AcroMetrix HIV-1 Low Control
AcroMetrix HIV-1 Mid Control
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
23-279-116 AcroMetrix HIV-1 Mid Control
23-279-115 AcroMetrix HIV-1 Low Control
23-279-117 AcroMetrix HIV-1 High Control
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-279-116 Supplier Thermo Scientific™ Supplier No. 964002
Add to Cart
Add to Cart
Identify increases in random or systematic error of nucleic acid test procedures for HIV-1 RNA with these controls, which are for In Vitro Diagnostic Use.
Assess the performance of molecular diagnostic test procedures, including the RNA extraction step, for the quantitative and qualitative determination of human immunodeficiency virus type-1 (HIV-1) RNA with the Thermo Scientific™ AcroMetrix™ HIV-1 Controls. These controls can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits and to evaluate and compare intralaboratory and interlaboratory assessments. The controls are for In Vitro Diagnostic Use.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description AcroMetrix HIV-1 Mid Control
Detectable Analytes HIV-1
Type HIV-1 Control
Packaging 5 x 1.2mL
Quantity 5 x 1.2 mL