Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AcroMetrix™ HIV-1/HCV/HBV Panels (RUO)
GSA_VA

Catalog No. 23279119
Change view
Click to view available options
Description:
AcroMetrix HBV Panel
AcroMetrix HCV-s Panel
AcroMetrix HIV-1 Panel (IU/mL)
AcroMetrix HIV-1 Panel (copies/mL)
Level:
0, 100, 1000, 10000, 100000, 1000000, 10000000 IU/mL
0, 100, 500, 5000, 50000, 500000, 5000000 copies/mL
100, 500, 5000, 50000, 500000, 5000000, 25000000 IU/mL
50, 500, 5000, 50000, 500000, 5000000, 50000000 IU/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Level
23-279-119 AcroMetrix HBV Panel 50, 500, 5000, 50000, 500000, 5000000, 50000000 IU/mL
23-279-113 AcroMetrix HIV-1 Panel (IU/mL) 0, 100, 1000, 10000, 100000, 1000000, 10000000 IU/mL
23-279-114 AcroMetrix HIV-1 Panel (copies/mL) 0, 100, 500, 5000, 50000, 500000, 5000000 copies/mL
23-279-123 AcroMetrix HCV-s Panel 100, 500, 5000, 50000, 500000, 5000000, 25000000 IU/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23-279-119 Supplier Thermo Scientific™ Supplier No. 950150
Add to Cart
Add to Cart

Assess the performance of molecular test procedures for HIV-1, HCV and HBV nucleic acids with these panels, which are for Research Use Only.

Assess the performance of molecular test procedures, including the extraction step, for the quantitative and qualitative determination of HIV-1, HCV and HBV nucleic acids with the Thermo Scientific™ AcroMetrix™ HIV-1/HCV/HBV Panels (RUO). These panels can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits and to evaluate and compare intralaboratory and interlaboratory assessments. The panels are for Research Use Only and are not for use in diagnostic procedures.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description AcroMetrix HBV Panel
Detectable Analytes HBV
Type HBV Panel
Level 50, 500, 5000, 50000, 500000, 5000000, 50000000 IU/mL
Packaging 7 x 1.2mL
Quantity 7 x 1.2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.