Learn More
Thermo Scientific™ AcroMetrix™ HPV Controls (RUO)
GSA_VA
Catalog No. 23279130
Change view
Click to view available options
Description:
AcroMetrix HPV Negative Control
AcroMetrix HPV-16 Genotype Control
AcroMetrix HPV-18 Genotype Control
AcroMetrix HPV-68 Genotype Control
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
23-279-130 AcroMetrix HPV-16 Genotype Control
23-279-131 AcroMetrix HPV-18 Genotype Control
23-279-132 AcroMetrix HPV-68 Genotype Control
23-279-133 AcroMetrix HPV Negative Control
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23-279-130 Supplier Thermo Scientific™ Supplier No. 950075
Add to Cart
Add to Cart
Assess the performance of nucleic acid test procedures for the determination of HPV DNA with these controls, which are for Research Use Only.
Assess the performance of nucleic acid test procedures for the qualitative determination of human papillomavirus DNA (HPV DNA) with the Thermo Scientific™ AcroMetrix™ HPV Controls (RUO). Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for Research Use Only and are not for use in diagnostic procedures.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description AcroMetrix HPV-16 Genotype Control
Type HPV Control
Packaging 5 x 4mL
Quantity 5 x 4 mL