Learn More
Thermo Scientific™ AcroMetrix™ HPV Controls (RUO)
Description
Assess the performance of nucleic acid test procedures for the qualitative determination of human papillomavirus DNA (HPV DNA) with the Thermo Scientific™ AcroMetrix™ HPV Controls (RUO). Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for Research Use Only and are not for use in diagnostic procedures.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | AcroMetrix HPV-18 Genotype Control |
| Type | HPV Control |
| Packaging | 5 x 4mL |
| Quantity | 5 x 4 mL |