Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AcroMetrix™ Transplant Virus Panels (RUO)

Catalog No. 23279155
Change view
Click to view available options
Description:
AcroMetrix Adenovirus Plasma Panel
AcroMetrix BKV Panel
AcroMetrix CMVtc Panel
AcroMetrix EBV Plasma Panel
AcroMetrix HSV-1 Plasma Panel
AcroMetrix HSV-2 Plasma Panel
AcroMetrix VZV Plasma Panel
Packaging:
5 x 0.5mL
5 x 0.8mL
Level:
100, 1000, 10000, 100000, 1000000 IU/mL
100, 1000, 10000, 100000, 1000000 copies/mL
1000, 10000, 100000, 1000000, 10000000 copies/mL
300, 3000, 30000, 300000, 3000000 IU/mL
500, 5000, 50000, 500000, 5000000 copies/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Packaging Level
23-279-155 AcroMetrix HSV-2 Plasma Panel 5 x 0.5mL 100, 1000, 10000, 100000, 1000000 copies/mL
23-279-158 AcroMetrix VZV Plasma Panel 5 x 0.5mL 100, 1000, 10000, 100000, 1000000 copies/mL
23-279-146 AcroMetrix EBV Plasma Panel 5 x 0.5mL 100, 1000, 10000, 100000, 1000000 IU/mL
23-279-141 AcroMetrix CMVtc Panel 5 x 0.8mL 300, 3000, 30000, 300000, 3000000 IU/mL
23-279-151 AcroMetrix HSV-1 Plasma Panel 5 x 0.5mL 100, 1000, 10000, 100000, 1000000 copies/mL
23-279-163 AcroMetrix Adenovirus Plasma Panel 5 x 0.5mL 1000, 10000, 100000, 1000000, 10000000 copies/mL
23-279-138 AcroMetrix BKV Panel 5 x 0.5mL 500, 5000, 50000, 500000, 5000000 copies/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 23-279-155 Supplier Thermo Scientific™ Supplier No. 954525
Add to Cart
Add to Cart

Assess the analytical performance of molecular test procedures for transplant viruses with these panels, which are for Research Use Only.

Assess the analytical performance of molecular test procedures, including the nucleic acid extraction step, for the quantitative and qualitative determination of viruses related to transplant disease with the Thermo Scientific™ AcroMetrix™ Transplant Virus Panels (RUO). These panels can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits, and to evaluate and compare intralaboratory and interlaboratory assessments. The panels are for Research Use Only and are not for use in diagnostic procedures.

TRUSTED_SUSTAINABILITY

Specifications

Description AcroMetrix HSV-2 Plasma Panel
Type Plasma Panel
Level 100, 1000, 10000, 100000, 1000000 copies/mL
Packaging 5 x 0.5mL
Quantity 5 x 0.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.