Learn More
Thermo Scientific™ AcroMetrix™ VZV Controls (RUO)
Catalog No. 23279159
Change view
Click to view available options
Description:
AcroMetrix VZV High CSF Control
AcroMetrix VZV High Plasma Control
AcroMetrix VZV Low CSF Control
Level:
High
Low
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Level
23-279-159 AcroMetrix VZV Low CSF Control Low
23-279-161 AcroMetrix VZV High Plasma Control High
23-279-160 AcroMetrix VZV High CSF Control High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-279-159 Supplier Thermo Scientific™ Supplier No. 954511
Add to Cart
Add to Cart
Identify increases in random or systematic error of nucleic acid test procedures for human VZV DNA with these controls, which are for Research Use Only.
Identify increases in random or systematic error of nucleic acid test procedures for the quantitative and qualitative determination of human VZV DNA with the Thermo Scientific™ AcroMetrix™ VZV Controls (RUO). Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for Research Use Only and are not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY
Specifications
Description AcroMetrix VZV Low CSF Control
Type VZV Control
Level Low
Packaging 5 x 0.5mL
Quantity 5 x 0.5 mL