Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva AcroPrep™ 24-well Filter Plates

Catalog No. 502110880 Shop All Cytiva Products
Encompass
Change view
Click to view available options
Quantity:
2/Pk.
8/Pk.
Membrane Type:
Omega
PP/PE
Supor
Pore Size:
0.1 μm
0.45 μm
0.8 μm
1.2 μm
30 to 40 μm
5 μm
MWCO:
1 kDa
10 kDa
100 kDa
3 kDa
30 kDa
50 kDa
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Membrane Type Pore Size MWCO
50-211-0880 2/Pk. Supor 0.1 μm -
50-211-0879 8/Pk. Supor 0.1 μm -
50-211-0882 2/Pk. Supor 0.45 μm -
50-211-0881 8/Pk. Supor 0.45 μm -
50-211-0884 2/Pk. Supor 0.8 μm -
50-211-0886 2/Pk. Supor 1.2 μm -
50-211-0885 8/Pk. Supor 1.2 μm -
50-211-0888 2/Pk. Supor 5 μm -
50-211-0887 8/Pk. Supor 5 μm -
50-211-0889 8/Pk. Omega - 1 kDa
50-211-0891 8/Pk. Omega - 3 kDa
50-211-0894 2/Pk. Omega - 10 kDa
50-211-0895 8/Pk. Omega - 30 kDa
50-211-0897 8/Pk. Omega - 50 kDa
50-211-0900 2/Pk. Omega - 100 kDa
50-211-0899 8/Pk. Omega - 100 kDa
50-211-0902 2/Pk. PP/PE 30 to 40 μm -
50-211-0901 8/Pk. PP/PE 30 to 40 μm -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 50-211-0880 Supplier Cytiva Supplier No. 97030
Add to Cart
Add to Cart

A one-and-done solution for filtration in a 24-well format: the AcroPrep™ 24-well filter plate range. For applications including protein purification, sterile filtration, general aqueous filtration, desalting, diafiltration, concentration, and fractionation of protein and nucleic acid samples.

Utilizing proprietary high-performance multi-layer filter membranes, plates offer time savings strong performance claims and streamlined workflow improvements. Available in 4 different types of membranes. Each plate comes with a V-shaped bottom collection plate and lid. Automation friendly. Vacuum compatible. Suitable for centrifuge.

 

Plates with Asymmetric Supor™ EKV Membrane:

A 0.65/0.2 μm asymmetric Supor EKV PES membrane, available with and without a dual-layer depth prefilter. The depth prefilter allows clarification of large cellular debris prior to filtration through the 0.65/0.2 μm EKV Supor membrane. Dual-layer clarification plate has reliable recovery of > 95% of extra cellular proteins. Single-layer AcroPrep 24-well filter plate contains 0.65/0.2 μm Supor EKV membrane, and is designed for applications where a sterile filtrate is required.

 

Plates with Supor Membrane:

The AcroPrep Supor plates have a variety of uses, in applications such as resin packing for sample preparation, viral filtration, or genome mapping, depending on the pore size chosen. Available with 0.1, 0.45, 0.8, 1.2 and 5 μm pore size ratings, these versatile filter plates can be confidently incorporated into a number of laboratory 24-well format workflows. The Supor polyethersulfone membrane enables fast filtration, has low protein binding properties and offers extensive drug and chemical compatibility. Each plate comes with a V-shaped bottom collection plate and lid Individually bagged and gamma irradiated. Automation friendly vacuum compatible suitable for centrifuge.

 

Plates with Omega Membrane:

The AcroPrep Omega plate for desalting, diafiltration, concentration or fractionation of protein and nucleic acid samples saves time in a single device and workflow step. The Omega ultrafiltration membrane is a polyethersulfone membrane specifically modified to minimize protein and nucleic acid binding. The Omega membrane works on the premise of size exclusion, where samples are fractionated by molecular weight. Larger molecules can be separated from smaller molecules by selecting the appropriate molecular weight cut-off (MWCO) and filtering the sample through these plates by either centrifugation, vacuum, or positive pressure, typically resulting in ≥ 90% recovery of target molecules.

 

Plates with PE/PP Membrane: 

The PE/PP plate offers a large pore size (30 to 40 μm) which is ideal for plant applications or as a cell strainer.

TRUSTED_SUSTAINABILITY

Specifications

Material Polypropylene
Pore Size 0.1 μm
Sterility Sterile
Filtration Area 1.6 cm2
Membrane Type Supor
Quantity 2/Pk.
Type Filter Plate with Membrane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.