Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acrosin Antibody (ACR-2), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500419H
Description
Acrosin Monoclonal antibody specifically detects Acrosin in Porcine, Bovine (Negative), Canine (Negative), Human (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Flow (Intracellular), CyTOFSpecifications
Acrosin | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Pig, Bovine (Negative), Canine (Negative), Human (Negative) | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry, Flow Cytometry (Intracellular Staining), CyTOF | |
ACR-2 | |
Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Flow (Intracellular), CyTOF-ready | |
acrosin, ACRS, preproacrosin, proacrosin | |
Acid extracts of boar spermatozoa were subjected to hydrophobic chromatography and the pooled fraction with reactivity to N-alpha benzoylarginine-4-nitroanilide was used for immunization. | |
0.1 mL | |
Signal Transduction | |
49 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction