Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Acrosin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561804
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561804 Supplier Invitrogen™ Supplier No. PA561804
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LMEARVDLID LDLCNSTQWY NGRVQPTNVC AGYPVGKIDT CQ Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

ACR is the major proteinase present in the acrosome of mature spermatozoa. It is a typical serine proteinase with trypsin-like specificity. It is stored in the acrosome in its precursor form, proacrosin. The active enzyme functions in the lysis of the zona pellucida, thus facilitating penetration of the sperm through the innermost glycoprotein layers of the ovum. The mRNA for proacrosin is synthesized only in the postmeiotic stages of spermatogenesis. In humans proacrosin first appears in the haploid spermatids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acrosin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACR
Gene Accession No. P10323
Gene Alias 53 kDa fucose-binding protein; Acr; Acro; Acrosin; Acrosin heavy chain; acrosin light and heavy chain prepropeptide; Acrosin light chain; acrosin precursor (EC 3.4.21.10); acrosin prepropeptide; ACRS; preproacrosin; proacrosin
Gene Symbols ACR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Acrosin. Recombinant protein control fragment (Product #RP-99045).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 49
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.