Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Acrosomal Vesicle Protein Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596930
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596930 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596930 Supplier Invitrogen™ Supplier No. PA596930
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QPNELSGSID HQTSVQQLPG EFFSLENPSD AEALYETSSG LNTLSEHGSS EHGSSKHTVA EHTSGEHAES EHASGEPAAT EHAEGEHTVG EQPSGEQPSG EHLSGEQPLS ELESGEQPSD EQPSGEHGSG EQPSGEQASG EQPSGEHASG EQASGAPISS TSTGTILNCY TCAYMNDQGK CLRGEGTCIT QNSQQCMLKK IFEGGKLQFM VQGCENMCPS MNLFSHGTRM QIICCRNQSF CNKI; Positive Samples: Mouse pancreas; Cellular Location: Cytoplasmic vesicle, acrosome, secretory vesicle.

This gene encodes a testis-specific, differentiation antigen, acrosomal vesicle protein 1, that arises within the acrosomal vesicle during spermatogenesis, and is associated with the acrosomal membranes and matrix of mature sperm. This gene consists of 4 exons and its alternative splicing generates multiple distinct transcripts, which encode protein isoforms ranging from 81 to 265 amino acids. The longest transcript is the most abundant, comprising 53-72% of the total acrosomal vesicle protein 1 messages; the second largest transcript comprises 15-32%; the third and the fourth largest transcripts account for 3.4-8.3% and 8.7-12.5%, respectively; and the remaining transcripts combined account for < 1% of the total acrosomal vesicle protein 1 message. It is suggested that phenomena of cryptic splicing and exon skipping occur within this gene. The acrosomal vesicle protein 1 may be involved in sperm-zona binding or penetration, and it is a potential contraceptive vaccine immunogen for humans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acrosomal Vesicle Protein
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ACRV1
Gene Accession No. P26436, P50289
Gene Alias acrosomal protein SP-10; Acrosomal vesicle protein 1; Acrv1; D11S4365; Msa63; MSA-63; rSP10; Sp10; SP-10; SPACA2; sperm membrane protein; sperm protein 10
Gene Symbols ACRV1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 22-265 of human ACRV1 (NP_0016031).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60353
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.