Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACRV1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501729170 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9170 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9170 Supplier Proteintech Group Inc Supplier No. 140401AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

One of the most frequent causes of man infertility is defective sperm acrosome. This damage can be detected using antibodies against intra-acrosomal proteins. Besides diagnostics of sperm pathology, monoclonal antibodies against intra-acrosomal proteins can be used for evaluation of the physiological state of sperm cells as well as for selection of a suitable method of fertilization in the laboratories of assisted reproduction.

Specifications

Antigen ACRV1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ACRV1
Gene Accession No. P26436
Gene Alias Acrosomal protein SP 10, acrosomal vesicle protein 1, ACRV1, D11S4365, SP 10, SPACA2
Gene Symbols ACRV1
Host Species Rabbit
Immunogen ACRV1 Fusion Protein Ag5109
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.