Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acrylamide/Bisacrylamide Solution (40%, 37.5:1), PROMO, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
1 L
Description
General Description
- Acrylamide/Bisacrylamide solution is used to to separate proteins
- The ratio of bisacrylamide to acrylamide modulates the porosity of the polyacrylamide gel
Application
- Acrylamide/Bisacrylamide Solution is used as a DNA sequencing gel promoting protein separation in the electrophoretic gels
- This solution is also applied for preparing gels using stock solutions with the most common cross-link ratios
- It has also been used as a precursor for polyacrylamide lid fabrication by photopatterning methods

Specifications
Specifications
Chemical Name or Material | Acrylamide/Bisacrylamide |
Physical Form | Liquid |
Color | Colorless |
Quantity | 500 mL |
MDL Number | MFCD00080848 |
UN Number | UN3426 |
Solubility Information | It is soluble in water. |
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction