Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrylamide Solution 1000 ppm in 80% Methanol, Reagents

Catalog No. 01336014
Click to view available options
Quantity:
500 mL

Acrylamide Solution 1000 ppm in 80% Methanol, Reagent, use for Acrylamide analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Acrylamide Solution 1000 ppm in 80% Methanol
Packaging Glass Bottle
CAS 67-56-1, 7732-18-5, 7732-18-5
Specific Gravity 0.8347
Physical Form Liquid
Color Colorless
Boiling Point 68°C
Solubility Completely miscible
Type Conductivity Standard
Quantity 500 mL
Grade Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.