Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrylamide ultra sequencing gel, 4%, for biochemistry, ready-to-use solution, Thermo Scientific™

Catalog No. AC330001000
Click to view available options
Quantity:
100 mL

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-06-1, 110-26-9, 57-13-6
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
Synonym Ethylenecarboxamide, 2-Propenamide
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Physical Form Liquid
CAS Min % 3.8, 95.8
CAS Max % 4.2, 96.2
Composition acrylamide 4X: 3.8% to 4.2%, methylene-bis acrylamide: 0.2%, urea: 36%
Color Colorless
pH 8.3 (Buffer TBE)
Quantity 100 mL
Beilstein 02, 400
Merck Index 15, 120
Formula Weight 71.08
Grade Biochemical
Chemical Name or Material Acrylamide ultra sequencing gel, 4%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes skin irritation. Causes damage to organs through prolonged or repeated exposure. Harmful if inhaled. Harmful if swallowed. May cause genetic defects. Suspected of damaging fertility or the unborn child. Toxic in contact with skin. May cause an allergic skin reaction. May cause cancer.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Gently wash with plenty of soap and water. Wear eye protection/face protection. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.