Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrylamide ultra sequencing gel, 8%, for biochemistry, ready-to-use solution, Thermo Scientific™

Catalog No. AC330021000
Click to view available options
Quantity:
100 mL

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-06-1, 110-26-9, 57-13-6
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
Synonym Ethylenecarboxamide, 2-Propenamide
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Physical Form Liquid
CAS Min % 3.8, 95.8
CAS Max % 4.2, 96.2
Composition acrylamide 4X: 7.5% to 8.5%, methylene-bis-acrylamide: 0.4%, urea: 45%
Color Colorless
pH 8.3 (Buffer TBE)
Quantity 100 mL
Packaging Glass Bottle
Beilstein 02, 400
Merck Index 15, 120
Formula Weight 71.08
Grade Biochemical
Chemical Name or Material Acrylamide ultra sequencing gel, 8%
Show More Show Less
Hazard Statement GHS H Statement:
May cause genetic defects.
May cause cancer.
Suspected of damaging fertility.
May cause an allergic skin reaction.
Harmful if swallowed.
Causes serious eye irritation.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.