Learn More
Eisco™ Acrylic Rectangular Block
Click to view available options
Dimensions (L x W x H):
75 x 50 x 18 mm
100 x 75 x 18 mm
115 x 65 x 18 mm
125 x 65 x 18 mm
Description
- Index of refraction plate
Specifications
Specifications
Product Type | Index of Refraction Plate |
Lens Material | Acrylic |
For Use With (Application) | Studying the properties of light (optics) in a kinesthetically and visually effective manner |
Dimensions (L x W x H) | 75 x 50 x 18 mm |