Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Acrylonitrile

Catalog No. 110213100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
5 mL
100 mL
500 mL
1 L
2 L
4 L
18 L

Catalog No. 11-021-3100ML

Supplier: Sigma Aldrich 110213100ML

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Acrylonitrile

Specifications

CAS 107-13-1
Melting Point -83°C
Boiling Point 77°C
Molecular Formula C3H3N
Refractive Index n20/D 1.391 (literature)
Linear Formula CH2=CHCN
Quantity 100 mL
MDL Number MFCD00001927
Synonym Vinyl cyanide
Molecular Weight (g/mol) 53.06
Percent Purity ≥99%
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.