Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrylonitrile, 99+%, stab. with ca 40ppm 4-methoxyphenol

Catalog No. p-7020934
Click to view available options
Quantity:
100 mL
500 mL
2500 mL

CAS: 107-13-1 | C3H3N | 53.064 g/mol

Acrylonitrile acts as a monomer used in the production of polyacrlonitrile plastics, adhesives and synthetic rubber. It is also used for the synthesis of copolymers such as styrene-acrylonitrile, acrylonitrile butadiene styrene, acrylonitrile styrene acrylate. It is involved in the preparation of adiponitrile by dimerization, which finds application in polyamide synthesis. It acts as a precursor to acrylamide and acrylic acid. It is used as dienophile in Diels-Alder reactions. It acts as a chemical intermediate in the synthesis of antioxidants, active pharmaceutical ingredients, dyes, and surface-active agents. Further, it is utilized for the preparation of acrylic fibers, resins and nitrile rubbers. In addition to this, it serves as a fumigant for food commodities, flour milling, and bakery food processing equipment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acrylonitrile acts as a monomer used in the production of polyacrlonitrile plastics, adhesives and synthetic rubber. It is also used for the synthesis of copolymers such as styrene-acrylonitrile, acrylonitrile butadiene styrene, acrylonitrile styrene acrylate. It is involved in the preparation of adiponitrile by dimerization, which finds application in polyamide synthesis. It acts as a precursor to acrylamide and acrylic acid. It is used as dienophile in Diels-Alder reactions. It acts as a chemical intermediate in the synthesis of antioxidants, active pharmaceutical ingredients, dyes, and surface-active agents. Further, it is utilized for the preparation of acrylic fibers, resins and nitrile rubbers. In addition to this, it serves as a fumigant for food commodities, flour milling, and bakery food processing equipment.

Solubility
Miscible with isopropanol, ethanol, ether, acetone, water, carbon tetrachloride, ethyl acetate, toluene and benzene.

Notes
Light sensitive. Incompatible with oxidizing agents, acids, alkalis, metals, bromine, peroxides and copper.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-13-1
Molecular Formula C3H3N
Molecular Weight (g/mol) 53.064
MDL Number MFCD00001927
InChI Key NLHHRLWOUZZQLW-UHFFFAOYSA-N
Synonym acrylonitrile, 2-propenenitrile, vinyl cyanide, propenenitrile, cyanoethylene, carbacryl, fumigrain, acritet, acrylon, ventox
PubChem CID 7855
ChEBI CHEBI:28217
IUPAC Name prop-2-enenitrile
SMILES C=CC#N

Specifications

Melting Point -84°C
Density 0.806
Boiling Point 77°C to 78°C
Flash Point 0°C (32°F)
Odor Pungent
Assay Percent Range 99+%
Refractive Index 1.391
Linear Formula CH2=CHCN
Quantity 2500 mL
UN Number UN1093
Beilstein 605310
Sensitivity Light sensitive
Merck Index 14,131
Solubility Information Miscible with isopropanol,ethanol,ether,acetone,water,carbon tetrachloride,ethyl acetate,toluene and benzene.
Formula Weight 53.06
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Acrylonitrile, stabilized with ≈40ppm 4-methoxyphenol
Show More Show Less
Hazard Category H225-H301+H311+H331-H315-H317-H318-H335-H350-H361
Hazard Statement GHS H Statement
H225-H301-H311-H331-H350-H318-H315-H317-H335
Highly flammable liquid and vapour.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
Causes serious eye damage.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280g-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P311-P312-P330-P333+P313-P36
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: ACRYLONITRILE, STABILIZED
EINECSNumber 203-466-5
RTECSNumber AT5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.