Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACSBG1 Mouse anti-Human, Clone: OTI2B6, liquid, TrueMAB™
SDP

Catalog No. 501672321
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-2321 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-2321 Supplier OriGene Supplier No. TA501176S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The protein encoded by this gene possesses long-chain acyl-CoA synthetase activity. It is thought to play a central role in brain very long-chain fatty acids metabolism and myelinogenesis.

Specifications

Antigen ACSBG1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2B6
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ACSBG1
Gene Accession No. Q96GR2
Gene Alias BG, BG1, BGM, GR-LACS, LPD
Gene Symbols ACSBG1
Host Species Mouse
Immunogen Full length human recombit protein of human ACSBG1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23205
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.