Learn More
ACSBG1 Mouse anti-Human, Clone: OTI2B6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501176

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene possesses long-chain acyl-CoA synthetase activity. It is thought to play a central role in brain very long-chain fatty acids metabolism and myelinogenesis.Specifications
ACSBG1 | |
Monoclonal | |
Unconjugated | |
ACSBG1 | |
BG, BG1, BGM, GR-LACS, LPD | |
Mouse | |
Affinity Chromatography | |
RUO | |
23205 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2B6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96GR2 | |
ACSBG1 | |
Full length human recombit protein of human ACSBG1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.