Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACSBG1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559659
Description
Immunogen sequence: ANVIMVDTQK QLEKILKIWK QLPHLKAVVI YKEPPPNKMA NVYTMEEFME LGNEVPEEAL DAIIDTQQPN QCCVLV Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
The protein encoded by this gene possesses long-chain acyl-CoA synthetase activity. It is thought to play a central role in brain very long-chain fatty acids metabolism and myelinogenesis.
Specifications
ACSBG1 | |
Polyclonal | |
Unconjugated | |
ACSBG1 | |
ACSBG1; Acyl-CoA synthetase bubblegum family member 1; BG; BG1; BG1 protein; BGM; bubblegum; E230019G03Rik; Gonadotropin-regulated long chain acyl CoA synthetase; gonadotropin-regulated long chain acyl-CoA synthetase; Grlacs; GR-LACS; hBG1; KIAA0631; Lipidosin; lipidosis-related protein lipidosin; lipodisin; long-chain-fatty-acid--CoA ligase ACSBG1; LPD; mBG1; mKIAA0631; R75185; very long-chain acyl-CoA synthetase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23205 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96GR2 | |
ACSBG1 | |
Recombinant protein corresponding to Human ACSBG1. Recombinant protein control fragment (Product #RP-98019). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction