Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSBG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570505
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570505 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570505 Supplier Invitrogen™ Supplier No. PA570505
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 86%; Guinea Pig: 86%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 93%.

The protein encoded by this gene possesses long-chain acyl-CoA synthetase activity. It is thought to play a central role in brain very long-chain fatty acids metabolism and myelinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSBG1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ACSBG1
Gene Accession No. Q96GR2
Gene Alias ACSBG1; Acyl-CoA synthetase bubblegum family member 1; BG; BG1; BG1 protein; BGM; bubblegum; E230019G03Rik; Gonadotropin-regulated long chain acyl CoA synthetase; gonadotropin-regulated long chain acyl-CoA synthetase; Grlacs; GR-LACS; hBG1; KIAA0631; Lipidosin; lipidosis-related protein lipidosin; lipodisin; long-chain-fatty-acid--CoA ligase ACSBG1; LPD; mBG1; mKIAA0631; R75185; very long-chain acyl-CoA synthetase
Gene Symbols ACSBG1
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human ACSBG1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23205
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.