Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSBG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562292
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562292 Supplier Invitrogen™ Supplier No. PA562292
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IYATNSAEAC QYVITHAKVN ILLVENDQQL QKILSIPQSS LEPLKAIIQY RLPMKKNNNL YSWDDFMELG RSIPDTQLEQ VIESQKANQC AVL Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 72%.

Catalyzes the conversion of fatty acids such as long chain and very long-chain fatty acids to their active form acyl-CoAs for both synthesis of cellular lipids, and degradation via beta-oxidation. Can activate diverse saturated, monosaturated and polyunsaturated fatty acids (PubMed:16371355, PubMed:16762313). Has increased ability to activate oleic and linoleic acid (PubMed:16371355). May play a role in spermatogenesis (PubMed:15685348). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSBG2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACSBG2
Gene Accession No. Q5FVE4
Gene Alias ACSBG2; Acyl-CoA synthetase bubblegum family member 2; Arachidonate--CoA ligase ACSBG2; BGR; BRGL; bubblegum-related protein; Long-chain-fatty-acid--CoA ligase ACSBG2; PRTDNY3; PRTD-NY3; testicular tissue protein Li 8; UNQ2443/PRO5005
Gene Symbols ACSBG2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACSBG2. Recombinant protein control fragment (Product #RP-99079).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81616
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.