Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ACSF
GSA_VAGREENER_CHOICESDP

Catalog No. 352525ML Shop All Novus Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-252-5ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 35-252-5ML Supplier R&D Systems™ Supplier No. 3525/25ML

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Artificial cerebrospinal fluid

Artificial cerebrospinal fluid (aCSF) commonly used when sampling from brain interstitial fluid. Closely matches the electrolyte concentrations of CSF and is prepared from high purity water and analytical grade reagents. Microfiltered and sterile. Final ion concentrations (in mM): Na+ 150; K+ 3.0; Ca2+ 1.4; Mg2+ 0.8; P 1.0; Cl- 155.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Quantity 25 mL
Product Line Tocris
Purity >98%
Chemical Name or Material ACSF
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.