Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACSF2 Antibody (18B4), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259501AF532
This item is not returnable.
View return policy
Description
ACSF2 Monoclonal antibody specifically detects ACSF2 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
ACSF2 | |
Monoclonal | |
Alexa Fluor 532 | |
ACSMW, acyl-CoA synthetase family member 2, AVYV493, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.26, mitochondrial, PPARG binding, long chain fatty acid acyl Co-A ligase like | |
Recombinant human ACSF2 (42-615aa) purified from E. coli | |
0.1 mL | |
Lipid and Metabolism | |
80221 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunofluorescence | |
18B4 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction