Learn More
Invitrogen™ ACSF2 Monoclonal Antibody (OTI6E4)

Description
ACSF2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
Specifications
| Antigen | ACSF2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI6E4 |
| Concentration | 0.53 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Acsf2 |
| Gene Accession No. | Q96CM8 |
| Gene Alias | ACSF2; ACSMW; acyl-CoA synthetase family member 2; acyl-CoA synthetase family member 2, mitochondrial; AVYV493; Medium-chain acyl-CoA ligase ACSF2, mitochondrial; PPARG binding, long chain fatty acid acyl Co-A ligase like; UNQ493/PRO1009 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.