Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACSF2 Monoclonal Antibody (OTI6E4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504711S


Description
ACSF2 Monoclonal antibody specifically detects ACSF2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)Specifications
ACSF2 | |
Monoclonal | |
0.53 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q86W50 | |
Mouse | |
Affinity chromatography | |
RUO | |
79066 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
OTI6E4 | |
Unconjugated | |
METTL16 | |
ACSF2; ACSMW; acyl-CoA synthetase family member 2; acyl-CoA synthetase family member 2, mitochondrial; AVYV493; Medium-chain acyl-CoA ligase ACSF2, mitochondrial; PPARG binding, long chain fatty acid acyl Co-A ligase like; UNQ493/PRO1009 | |
Full length human recombinant protein of human METT10D produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction