Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACSL1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11099585AF488
Description
ACSL1 Polyclonal specifically detects ACSL1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| ACSL1 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| P33121 | |
| ACSL1 | |
| Synthetic peptide made to an N-terminal portion of the human ACSL1 protein (within residues 1-100). [Swiss-Prot# P33121] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Primate | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ACS1LACS 2, Acyl-CoA synthetase 1, acyl-CoA synthetase long-chain family member 1, EC 6.2.1, FACL1EC 6.2.1.3, fatty-acid-Coenzyme A ligase, long-chain 1, LACS 1, LACSlong-chain 2, lignoceroyl-CoA synthase, Long-chain acyl-CoA synthetase 1, Long-chain acyl-CoA synthetase 2, Long-chain fatty acid-CoA ligase 2, long-chain fatty-acid-coenzyme A ligase 1, long-chain-fatty-acid--CoA ligase 1, Palmitoyl-CoA ligase 1, Palmitoyl-CoA ligase 2, paltimoyl-CoA ligase 1 | |
| Rabbit | |
| 78 kDa | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism, Signal Transduction | |
| 2180 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction