Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSL1 Recombinant Rabbit Monoclonal Antibody (HL1530)
GREENER_CHOICE

Catalog No. PIMA547083
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547083 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547083 Supplier Invitrogen™ Supplier No. MA547083
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSL1
Applications Western Blot
Classification Recombinant Monoclonal
Clone HL1530
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ACSL1
Gene Accession No. P18163, P33121, P41216
Gene Alias Acas; Acas1; ACS; Acs1; ACSL1; Acsl2; Acyl CoA synthetase, long chain; acyl-CoA synthetase 1; acyl-CoA synthetase long chain family member 1; acyl-CoA synthetase long-chain family member 1; Arachidonate--CoA ligase; COAA; FACL1; Facl2; FACS; fatty acid Coenzyme A ligase, long chain 2; fatty-acid-Coenzyme A ligase long chain 2; fatty-acid-Coenzyme A ligase, long-chain 1; fatty-acid-Coenzyme A ligase, long-chain 2; L LACS2; LACS; LACS 1; LACS 2; LACS1; LACS2; lignoceroyl-CoA synthase; long-chain acyl-CoA synthetase 1; Long-chain acyl-CoA synthetase 2; long-chain fatty acid-CoA ligase 2; long-chain fatty-acid-coenzyme A ligase 1; long-chain-fatty-acid--CoA ligase 1; Long-chain-fatty-acid--CoA ligase, liver isozyme; OTTHUMP00000218457; OTTHUMP00000218460; OTTHUMP00000218462; OTTHUMP00000218465; palmitoyl-CoA ligase; Palmitoyl-CoA ligase 1; palmitoyl-CoA ligase 2; paltimoyl-CoA ligase 1; Phytanate--CoA ligase
Gene Symbols ACSL1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ACSL1. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14081, 2180, 25288
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.