Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582448
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582448 Supplier Invitrogen™ Supplier No. PA582448
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IIEQGCFAYS MVIVPLYDTL GNEAITYIVN KAELSLVFVD KPEKAKLLLE GVENKLIPGL KIIVVMDAYG SELVERGQRC GVEVTSMKAM EDLGRANRRK PKPPAPEDLA VICFTSGTTG NPKGAMVTHR N Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 81%.

The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSL1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACSL1
Gene Accession No. P33121
Gene Alias Acas; Acas1; ACS; Acs1; ACSL1; Acsl2; Acyl CoA synthetase, long chain; acyl-CoA synthetase 1; acyl-CoA synthetase long chain family member 1; acyl-CoA synthetase long-chain family member 1; Arachidonate--CoA ligase; COAA; FACL1; Facl2; FACS; fatty acid Coenzyme A ligase, long chain 2; fatty-acid-Coenzyme A ligase long chain 2; fatty-acid-Coenzyme A ligase, long-chain 1; fatty-acid-Coenzyme A ligase, long-chain 2; L LACS2; LACS; LACS 1; LACS 2; LACS1; LACS2; lignoceroyl-CoA synthase; long-chain acyl-CoA synthetase 1; Long-chain acyl-CoA synthetase 2; long-chain fatty acid-CoA ligase 2; long-chain fatty-acid-coenzyme A ligase 1; long-chain-fatty-acid--CoA ligase 1; Long-chain-fatty-acid--CoA ligase, liver isozyme; OTTHUMP00000218457; OTTHUMP00000218460; OTTHUMP00000218462; OTTHUMP00000218465; palmitoyl-CoA ligase; Palmitoyl-CoA ligase 1; palmitoyl-CoA ligase 2; paltimoyl-CoA ligase 1; Phytanate--CoA ligase
Gene Symbols ACSL1
Host Species Rabbit
Immunogen Recombinant protein fragment of Human ACSL1 (Product #RP-89594)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2180
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.