Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542883
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542883 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542883 Supplier Invitrogen™ Supplier No. PA542883
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LDGLASVLYP GCDTLDKVFT YAKNKFKNKR LLGTREVLNE EDEVQPNGKI Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 93%.

An initial reaction in fatty acid metabolism in eukaryotic cells is activation of fatty acids catalyzed by acyl-CoA synthetase. FACL3 (fatty acid CoA ligase, long-chain 3) is identified as member of the acyl-CoA synthetase (ACS) family by PCR of rat brain cDNAs using primers based on the conserved region of the ACS protein. The 720-amino acid rat protein preferentially utilizes myristate, laurate, arachidonate, and eicosapentaenoate, and is expressed primarily in brain. The predicted 720-amino acid FACL3 human protein is 92% identical to that of rat.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSL3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ACSL3
Gene Accession No. O95573
Gene Alias 2610510B12Rik; Acs3; Acsl3; acyl-CoA synthetase long chain family member 3; acyl-CoA synthetase long-chain family member 3; Arachidonate--CoA ligase; Brain acyl-CoA synthetase II; C85929; FACL3; fatty acid Coenzyme A ligase, long chain 3; fatty-acid-Coenzyme A ligase, long-chain 3; LACS 3; LACS3; lignoceroyl-CoA synthase; long-chain acyl-CoA synthetase 3; Long-chain-fatty-acid--CoA ligase 3; Pro2194
Gene Symbols ACSL3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ACSL3 (aa 74-123).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2181
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.