Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582427
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582427 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582427 Supplier Invitrogen™ Supplier No. PA582427
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MYNFQLVTLY ATLGGPAIVH ALNETEVTNI ITSKELLQTK LKDIVSLVPR LRHIITVDGK PPTWSEFPKG IIVHTMAAVE ALGAKASMEN QPHSKPLPSD IAVIM Highest antigen sequence indentity to the following orthologs: Rat - 90%, Mouse - 91%.

An initial reaction in fatty acid metabolism in eukaryotic cells is activation of fatty acids catalyzed by acyl-CoA synthetase. FACL3 (fatty acid CoA ligase, long-chain 3) is identified as member of the acyl-CoA synthetase (ACS) family by PCR of rat brain cDNAs using primers based on the conserved region of the ACS protein. The 720-amino acid rat protein preferentially utilizes myristate, laurate, arachidonate, and eicosapentaenoate, and is expressed primarily in brain. The predicted 720-amino acid FACL3 human protein is 92% identical to that of rat.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSL3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACSL3
Gene Accession No. O95573
Gene Alias 2610510B12Rik; Acs3; Acsl3; acyl-CoA synthetase long chain family member 3; acyl-CoA synthetase long-chain family member 3; Arachidonate--CoA ligase; Brain acyl-CoA synthetase II; C85929; FACL3; fatty acid Coenzyme A ligase, long chain 3; fatty-acid-Coenzyme A ligase, long-chain 3; LACS 3; LACS3; lignoceroyl-CoA synthase; long-chain acyl-CoA synthetase 3; Long-chain-fatty-acid--CoA ligase 3; Pro2194
Gene Symbols ACSL3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACSL3. Recombinant protein control fragment (Product #RP-89591).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2181
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.