Learn More
Invitrogen™ ACSL3 Polyclonal Antibody

Description
Positive Samples: HeLa, Mouse kidney; Cellular Location: Endoplasmic reticulum membrane, Microsome membrane, Mitochondrion outer membrane, Peroxisome membrane, Single-pass type III membrane protein Immunogen sequence: CEMENEVLKV LSEAAISASL E.
Specifications
Specifications
| Antigen | ACSL3 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.53 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | ACSL3 |
| Gene Accession No. | O95573, Q63151, Q9CZW4 |
| Gene Alias | 2610510B12Rik; Acs3; Acsl3; acyl-CoA synthetase long chain family member 3; acyl-CoA synthetase long-chain family member 3; Arachidonate--CoA ligase; Brain acyl-CoA synthetase II; C85929; FACL3; fatty acid Coenzyme A ligase, long chain 3; fatty-acid-Coenzyme A ligase, long-chain 3; LACS 3; LACS3; lignoceroyl-CoA synthase; long-chain acyl-CoA synthetase 3; Long-chain-fatty-acid--CoA ligase 3; Pro2194 |
| Gene Symbols | ACSL3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.