Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA512225
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA512225 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA512225 Supplier Invitrogen™ Supplier No. PA512225
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACSL4 Polyclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme preferentially utilizes arachidonate as substrate. The absence of this enzyme may contribute to the mental retardation or Alport syndrome. Alternative splicing of this gene generates 2 transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSL4
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Acsl4
Gene Accession No. O60488
Gene Alias 9430020A05Rik; Acs4; ACSL4; acyl-CoA synthetase 4; acyl-CoA synthetase long-chain family member 4; Arachidonate--CoA ligase; AU018108; Facl4; fatty acid Coenzyme A ligase, long chain 4; fatty acid-Coenzyme A ligase long chain 4; fatty acid-Coenzyme A ligase, long chain 4; fatty-acid-Coenzyme A ligase, long-chain 4; L MRX63; LACS 4; Lacs4; lignoceroyl-CoA synthase; long-chain acyl-CoA synthetase 4; long-chain fatty-acid-Coenzyme A ligase 4; long-chain-fatty-acid--CoA ligase 4; mACS4; MRX63; MRX68
Gene Symbols Acsl4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 236-267 amino acids from the central region of human ACSL4 (FACL4).
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2182
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.